Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isonipecotamide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
39546-32-2
C6H12N2O
128.17
MFCD00038012
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
isonipecotamide,4-piperidinecarboxamide,hexahydroisonicotinamide,4-carbamoylpiperidine,isonipectoamide,piperidine-4-carboxylic acid amide,4-piperdinecarboxamide,piperidin-4-carboxamid,iso nipecotamide,iso-nipecotamide
3772
piperidine-4-carboxamide
C1CNCCC1C(=O)N
39546-32-2
C6H12N2O
128.17
MFCD00038012
DPBWFNDFMCCGGJ-UHFFFAOYSA-N
isonipecotamide,4-piperidinecarboxamide,hexahydroisonicotinamide,4-carbamoylpiperidine,isonipectoamide,piperidine-4-carboxylic acid amide,4-piperdinecarboxamide,piperidin-4-carboxamid,iso nipecotamide,iso-nipecotamide
3772
piperidine-4-carboxamide
C1CNCCC1C(=O)N
Melting Point 146°C to 150°C
Infrared Spectrum Authentic
Quantity 500g
Water 2% max. (K.F.)
Loss on Drying 2% max. (70°C)
Color White to Yellow
Packaging Glass bottle
Sulfated Ash 0.2% max.
Assay Percent Range 98%
Solubility Information Solubility in water: soluble
Formula Weight 128.17
Physical Form Crystalline Powder
Percent Purity ≥97.5% (on dry substance)
Chemical Name or Material Isonipecotamide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning