Alfa Aesar™ Isopentyl 3-(methylthio)propionate, 98%

Manufacturer: Alfa Aesar™ B2231322

Quantity 100g

 View more versions of this product

Catalog No. AAB2231322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 93762-35-7
    Molecular Formula CH3SCH2CH2CO2CH2CH2CH(CH3)2
    Formula Weight 190.31
    MDL Number MFCD00053101
    Beilstein 7914388
    Synonym Isoamyl 3-(methylthio)propionate; 3-Methylbutyl 3-(methylthio)propionate
    Chemical Name or Material Isopentyl 3-(methylthio)propionate

    Specifications

    Formula Weight 190.31
    Molecular Formula CH3SCH2CH2CO2CH2CH2CH(CH3)2
    MDL Number MFCD00053101
    Synonym Isoamyl 3-(methylthio)propionate; 3-Methylbutyl 3-(methylthio)propionate
    Boiling Point 222°C
    Flash Point 115°C (239°F)
    Quantity 100g
    CAS 93762-35-7
    Beilstein 7914388
    Chemical Name or Material Isopentyl 3-(methylthio)propionate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit