Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isopentyl 3-(methylthio)propionate, 98%

Click to view available options
Quantity:
5g
25g
100g
93762-35-7
CH3SCH2CH2CO2CH2CH2CH(CH3)2
MFCD00053101
isopentyl 3-methylthio propionate,isoamyl 3-methylthio propionate,3-methylbutyl 3-methylsulfanyl propanoate,3-methylbutyl 3-methylthio propanoate,3-methylbutyl 3-methylthio propionate
93762-35-7
CH3SCH2CH2CO2CH2CH2CH(CH3)2
MFCD00053101
isopentyl 3-methylthio propionate,isoamyl 3-methylthio propionate,3-methylbutyl 3-methylsulfanyl propanoate,3-methylbutyl 3-methylthio propanoate,3-methylbutyl 3-methylthio propionate
Flash Point 115°C (239°F)
Refractive Index 1.4585
Density 0.976
Boiling Point 222°C
Quantity 5g
Beilstein 7914388
Formula Weight 190.31
Percent Purity 98%
Chemical Name or Material Isopentyl 3-(methylthio)propionate

EINECSNumber : 297-748-5

TSCA : No

Recommended Storage : Ambient temperatures