Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Isopentyl 3-(methylthio)propionate, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Density | 0.976 |
Refractive Index | 1.4585 |
Quantity | 25g |
Boiling Point | 222°C |
Flash Point | 115°C (239°F) |
Beilstein | 7914388 |
Formula Weight | 190.31 |
Percent Purity | 98% |
Chemical Name or Material | Isopentyl 3-(methylthio)propionate |
Safety and Handling
EINECSNumber : 297-748-5
TSCA : No
Recommended Storage : Ambient temperatures