Alfa Aesar™ Isopentyl isobutyrate, 98%

Manufacturer: Alfa Aesar™ B2487914

Quantity 25g

 View more versions of this product

Catalog No. AAB2487914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2050-01-3
    Molecular Formula C9H18O2
    Linear Formula (CH3)2CHCH2CH2O2CCH(CH3)2
    Formula Weight 158.24
    MDL Number MFCD00053719
    UN Number UN2620
    Synonym Isobutyric acid isoamyl ester
    Chemical Name or Material Isopentyl isobutyrate

    Specifications

    Formula Weight 158.24
    TSCA TSCA
    Molecular Formula C9H18O2
    Linear Formula (CH3)2CHCH2CH2O2CCH(CH3)2
    MDL Number MFCD00053719
    UN Number UN2620
    Synonym Isobutyric acid isoamyl ester
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Boiling Point 162°C
    Flash Point 56°C (133°F)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL BUTYRATES
    Quantity 25g
    CAS 2050-01-3
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Isopentyl isobutyrate
    Assay 98%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit