Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Isopentyl isobutyrate, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Density | 0.857 |
Refractive Index | 1.407 |
Flash Point | 56°C (133°F) |
Quantity | 500g |
Boiling Point | 162°C |
Linear Formula | (CH3)2CHCH2CH2O2CCH(CH3)2 |
UN Number | UN2620 |
Formula Weight | 158.24 |
Percent Purity | 98% |
Chemical Name or Material | Isopentyl isobutyrate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMYL BUTYRATES
EINECSNumber : 218-078-1
TSCA : Yes
Recommended Storage : Ambient temperatures