Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorondiamine, 99+%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
2855-13-2
C10H22N2
170.3
MFCD00019397
RNLHGQLZWXBQNY-UHFFFAOYSA-N
isophorone diamine,isophoronediamine,3-aminomethyl-3,5,5-trimethylcyclohexylamine,3-aminomethyl-3,5,5-trimethylcyclohexanamine,cyclohexanemethanamine, 5-amino-1,3,3-trimethyl,ipda,ccris 6680,5-amino-1,3,3-trimethylcyclohexanemethylamine,cyclohexanemethylamine, 5-amino-1,3,3-trimethyl,1-amino-3-aminomethyl-3,5,5-trimethylcyclohexane
17857
3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
CC1(CC(CC(C1)(C)CN)N)C
2855-13-2
C10H22N2
170.3
MFCD00019397
RNLHGQLZWXBQNY-UHFFFAOYSA-N
isophorone diamine,isophoronediamine,3-aminomethyl-3,5,5-trimethylcyclohexylamine,3-aminomethyl-3,5,5-trimethylcyclohexanamine,cyclohexanemethanamine, 5-amino-1,3,3-trimethyl,ipda,ccris 6680,5-amino-1,3,3-trimethylcyclohexanemethylamine,cyclohexanemethylamine, 5-amino-1,3,3-trimethyl,1-amino-3-aminomethyl-3,5,5-trimethylcyclohexane
17857
3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
CC1(CC(CC(C1)(C)CN)N)C
Density 0.9200g/mL
Refractive Index 1.4880 to 1.49
Melting Point 10°C
Boiling Point 247°C
Flash Point 117°C
Specific Gravity 0.92
Viscosity 18 mPa.s (20°C)
Packaging Glass bottle
Color Undesignated
Quantity 250mL
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Solubility Information Solubility in water: miscible with water
Formula Weight 170.3
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Isophorondiamine, 99+%
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-666-8

RTECSNumber : GV5020833

TSCA : TSCA