Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorone, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
1L
2.5L
Packaging:
Glass bottle
78-59-1
C9H14O
138.21
MFCD00001584
HJOVHMDZYOCNQW-UHFFFAOYSA-N
isophorone,isoacetophorone,isoforone,isoforon,izoforon,isooctopherone,3,5,5-trimethyl-2-cyclohexen-1-one,alpha-isophorone,2-cyclohexen-1-one, 3,5,5-trimethyl,1,1,3-trimethyl-3-cyclohexene-5-one
6544
CHEBI:34800
3,5,5-trimethylcyclohex-2-en-1-one
CC1=CC(=O)CC(C1)(C)C
78-59-1
C9H14O
138.21
MFCD00001584
HJOVHMDZYOCNQW-UHFFFAOYSA-N
isophorone,isoacetophorone,isoforone,isoforon,izoforon,isooctopherone,3,5,5-trimethyl-2-cyclohexen-1-one,alpha-isophorone,2-cyclohexen-1-one, 3,5,5-trimethyl,1,1,3-trimethyl-3-cyclohexene-5-one
6544
CHEBI:34800
3,5,5-trimethylcyclohex-2-en-1-one
CC1=CC(=O)CC(C1)(C)C
Viscosity 2.6 mPa.s (20°C)
Refractive Index 1.4770 to 1.4790
Specific Gravity 0.92
Boiling Point 213.0°C to 214.0°C
Melting Point -8.0°C
Density 0.9200g/mL
Packaging Glass bottle
Flash Point 84°C
Quantity 5mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Beilstein 07,65
Merck Index 15,5242
Solubility Information Solubility in water: 12g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone
Formula Weight 138.21
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Isophorone, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Suspected of causing cancer.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 201-126-0

RTECSNumber : GW7700000

TSCA : TSCA