Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophorone diisocyanate, mixture of isomers, 98%, Thermo Scientific Chemicals

Catalog Number p-7046268
Click to view available options
Quantity:
5 g
100 g
500 g
4098-71-9
C12H18N2O2
222.29
MFCD00064956
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
169132
CHEBI:53214
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
4098-71-9
C12H18N2O2
222.29
MFCD00064956
NIMLQBUJDJZYEJ-UHFFFAOYNA-N
isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
169132
CHEBI:53214
5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
TRUSTED_SUSTAINABILITY
Melting Point -60°C
Density 1.061
Boiling Point 153°C (10 mmHg)
Flash Point 163°C (325°F)
Refractive Index 1.484
Quantity 5 g
UN Number UN2290
Beilstein 2726467
Sensitivity Moisture sensitive
Solubility Information Miscible with esters,ketones,ethers,and aromatic and aliphatic hydrocarbons.
Formula Weight 222.29
Percent Purity 98%
Chemical Name or Material Isophorone diisocyanate, mixture of isomers
Show More Show Less
Hazard Category H315-H317-H319-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H315-H319-H317-H335-H204
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.
Precautionary Statement P260-P264b-P271-P272-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOPHORONE DIISOCYANATE
EINECSNumber 223-861-6
RTECSNumber NQ9370000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit