Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isophthalic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2kg
Packaging:
Glass bottle
Plastic bottle
121-91-5
C8H6O4
166.13
MFCD00002516
QQVIHTHCMHWDBS-UHFFFAOYSA-N
isophthalic acid,1,3-benzenedicarboxylic acid,m-phthalic acid,m-benzenedicarboxylic acid,acide isophtalique,kyselina isoftalova,iso-phthalic acid,acide isophtalique french,kyselina isoftalova czech,unii-x35216h9fj
8496
CHEBI:30802
benzene-1,3-dicarboxylic acid
C1=CC(=CC(=C1)C(=O)O)C(=O)O
121-91-5
C8H6O4
166.13
MFCD00002516
QQVIHTHCMHWDBS-UHFFFAOYSA-N
isophthalic acid,1,3-benzenedicarboxylic acid,m-phthalic acid,m-benzenedicarboxylic acid,acide isophtalique,kyselina isoftalova,iso-phthalic acid,acide isophtalique french,kyselina isoftalova czech,unii-x35216h9fj
8496
CHEBI:30802
benzene-1,3-dicarboxylic acid
C1=CC(=CC(=C1)C(=O)O)C(=O)O
Infrared Spectrum Authentic
Melting Point 341.0°C to 343.0°C
Color White
Quantity 5g
Packaging Glass bottle
Sulfated Ash 0.0015% max.
Assay Percent Range 99%
Linear Formula C6H4(CO2H)2
Beilstein 09, 832
Merck Index 15, 5243
Solubility Information Solubility in water: 0.01g/100mL (25 c). Other solubilities: freely soluble in alcohol and glacial acetic acid,practically insoluble in benzene and petroleum,ether
Formula Weight 166.13
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Isophthalic acid
Show More Show Less