Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Isopore™ Membrane Filters, 0.1 μm Available on GSA/VA Contract for Federal Government customers only.

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

$145.30 - $423.50

Specifications

Autoclavable Not Autoclavable
Color White
Material Polycarbonate
Pore Size 0.1 μm
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Bubble Point Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Bubble Point Price Quantity & Availability  
VCTP01300
View Documents
MilliporeSigma™
VCTP01300
13 mm 101 psi
Pack of 100 for $145.30
Add to cart
 
VCTP02500 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
VCTP02500
25 mm 101 psi
Pack of 100 for $153.50
Add to cart
 
VCTP04700
View Documents
MilliporeSigma™
VCTP04700
47 mm 101 psi
Pack of 100 for $272.00
Add to cart
 
VCTP14250
View Documents
MilliporeSigma™
VCTP14250
142 mm 7.0 bar, air with water
Pack for $423.50
Add to cart
 
Description

Description

Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.

The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Specifications

Specifications

Not Autoclavable
Polycarbonate
Non-sterile
Filter Membranes
White
0.1 μm
Screen filter
Hydrophilic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit