Learn More
Selleck Chemical LLC Isoprenaline (Isoproterenol) HCl S2566-1g
SDP
Supplier:  Selleck Chemical LLC S25661G
Encompass_Preferred
Isoprenaline (Isoproterenol) HCl is a non-selective beta-adrenergic receptor agonist used for the treatment of bradycardia and heart block
Catalog No. 50-311-2724