Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 70% (v/v), Ricca Chemical
SDP

Catalog Number p-2915847
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
67-63-0, 7732-18-5
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0, 7732-18-5
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
CAS Min % 29.0, 69.0
CAS Max % 31.0, 71.0
Appearance Colorless Liquid
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Molecular Weight (g/mol) 60.096
Percent Purity 68 to 72%
Quantity 500 mL
Packaging Natural Poly Bottle
Specific Gravity 0.874 to 0.882
Chemical Name or Material Isopropanol
ShelfLife 24 months
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit