Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropanol, 99.5+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
2.5L
10L
25L
Packaging:
Glass Bottle
Metal drum
Plastic drum
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
Assay Percent Range 99.5+%
Linear Formula (CH3)2CHOH
Merck Index 15, 5254
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Physical Form Liquid
Percent Purity ≥99.5%
Grade Extra Pure
Density 0.7850g/mL
Non_volatile Matter 0.001% max.
Packaging Glass Bottle
Refractive Index 1.3760 to 1.3780
Solubility Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Specific Gravity 0.785
Water 0.2% max.
Flash Point 12°C
Viscosity 2.27 mPa.s (20°C)
Boiling Point 81.0°C to 83.0°C
Color Undesignated
Melting Point -89.5°C
Quantity 2.5L
Chemical Name or Material Isopropanol
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger