Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl acetate, 99+%, for analysis, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
108-21-4
C5H10O2
102.13
MFCD00008877
JMMWKPVZQRWMSS-UHFFFAOYSA-N
isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
7915
propan-2-yl acetate
CC(C)OC(=O)C
108-21-4
C5H10O2
102.13
MFCD00008877
JMMWKPVZQRWMSS-UHFFFAOYSA-N
isopropyl acetate,2-propyl acetate,2-acetoxypropane,acetic acid, 1-methylethyl ester,isopropyl ethanoate,isopropylacetat,paracetat,isopropylacetaat,1-methylethyl acetate,acetic acid, isopropyl ester
7915
propan-2-yl acetate
CC(C)OC(=O)C
For analysis
Viscosity 0.49 mPa.s (25°C)
Quantity 2.5L
Boiling Point 88.8°C
Refractive Index 1.3760 to 1.3780
Infrared Spectrum Authentic
Water 0.1% max. (Coulometric)
Packaging Glass bottle
Specific Gravity 0.872
Residue after Evaporation 0.002% max.
Flash Point 4°C
Density 0.8720g/mL
Color Undesignated
Melting Point -73.0°C
Assay Percent Range 99+%
Linear Formula CH3CO2CH(CH3)2
Merck Index 15, 5251
Solubility Information Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 102.13
Physical Form Liquid
Percent Purity ≥99%
Grade Analysis
Chemical Name or Material Isopropyl acetate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger