Alfa Aesar™ Isopropyl acetoacetate, 98%

Manufacturer: Alfa Aesar™ A1190718

Quantity 50g

 View more versions of this product

Catalog No. AAA1190718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 542-08-5
    Molecular Formula C7H12O3
    Formula Weight 144.17
    MDL Number MFCD00043933
    Beilstein 1757046
    Merck Index 14,5206
    Synonym Acetoacetic acid isopropyl ester
    Chemical Name or Material Isopropyl acetoacetate

    Specifications

    Formula Weight 144.17
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H12O3
    Health Hazard 1 Warning
    Quantity 50g
    CAS 542-08-5
    MDL Number MFCD00043933
    Merck Index 14,5206
    Synonym Acetoacetic acid isopropyl ester
    Solubility Information Slightly soluble in water
    Melting Point −27°C
    TSCA TSCA
    Beilstein 1757046
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Flash Point 73°C (163°F)
    Boiling Point 185° to 187°C
    Chemical Name or Material Isopropyl acetoacetate
    Assay 98%

    Description

    Applications
    Isopropyl acetoacetate is used as an intermediate for the manufacturing of agrochemicals and pharmaceutical products such as isradipine.

    Solubility
    Slightly miscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit