Isopropyl Alchohol, 99%, World/GMP, ACS/USP/FCC/EP/BP/JP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 231WORLDDM55M

Packaging Metal drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-398

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-63-0
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula C3H8O
    Formula Weight 60.1
    Synonym 2-Propanol
    Chemical Name or Material Isopropyl Alcohol 99%

    Specifications

    Formula Weight 60.1
    Quantity 55 gal.
    Color Colorless
    Physical Form Liquid
    Packaging Metal drum
    Stability Stable under recommended storage conditions.
    Assay Percent Range 99%
    Molecular Formula C3H8O
    CAS 67-63-0
    Synonym 2-Propanol
    Description General use organic solvent
    Chemical Name or Material Isopropyl Alcohol 99%
    Assay 99%

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit