Isopropyl Alchohol, 99%, World/GMP, ACS/USP/FCC/EP/BP/JP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 231WORLDPL05M

Packaging Metal Pail
Quantity 5 gal.

 View more versions of this product

Catalog No. 16-101-401

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-63-0
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula C3H8O
    Formula Weight 60.1
    Synonym 2-Propanol
    Chemical Name or Material Isopropyl Alcohol 99%

    Specifications

    Boiling Point 83°C
    Melting Point −90°C
    Formula Weight 60.1
    Quantity 5 gal.
    Color Colorless
    Packaging Metal Pail
    Flash Point 12°C (53.6°F)
    UN Number 1219; Colorless
    Physical Form Liquid
    Assay Percent Range 99%
    Description General use organic solvent
    Stability Stable under recommended storage conditions.
    Molecular Formula C3H8O
    CAS 67-63-0
    Synonym 2-Propanol
    Chemical Name or Material Isopropyl Alcohol 99%
    Assay 99%

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit