Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alchohol, 99%, World/GMP, ACS/USP/FCC/EP/BP/JP Grade, PHARMCO-AAPER

Manufacturer:  Pharmco Products 231WORLDCS4L

Packaging Amber Gass Bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-101-397


  Check Availability
Add to cart

Alert:

Minimum Order Quantity: 5


Specifications

Description

Order Info

Minimum Order Quantity: 5

Description & Specifications

Chemical Identifiers

67-63-0
99%
60.1
Isopropyl Alcohol 99%
Liquid
C3H8O
2-Propanol

Specifications

67-63-0
99%
60.1
2-Propanol
Amber Gass Bottle
12°C (53.6°F)
Colorless
4L
99%
Liquid
C3H8O
1219
General use organic solvent
Stable under recommended storage conditions.
83°C
−90°C
Isopropyl Alcohol 99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit