Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 70%, GMP/USP/NF Grade, PHARMCO-AAPER

Manufacturer:  Pharmco Products 211000GMPDM55M

Packaging Metal drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-395


  Check Availability
Add to cart

Alert:

Minimum Order Quantity: 5


Specifications

Description

Order Info

Minimum Order Quantity: 5

Description & Specifications

Chemical Identifiers

67-63-0,7732-18-5
70%
60.1
Isopropyl Alcohol 70%
Liquid
C3H8O
2-Propanol

Specifications

67-63-0,7732-18-5
70%
60.1
2-Propanol
Metal drum
18.3°C (64.9° F)
−90°C
Isopropyl Alcohol 70%
Liquid
C3H8O
1219, Colorless, Clear
General use organic solvent
Stable under recommended storage conditions.
Clear colorless
55 gal.
70%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit