Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Anhydrous, Approx. 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O

I1275, 67-63-0, C3H8O

Spectrum™ Isopropyl Alcohol, Anhydrous is used when performing certain reactions where the presence of water can prevent a chemical reaction from happening, or form undesirable products. Spectrum anhydrous chemical solvents are packaged in convenient sizes featuring a double closure, which includes an easy-slip-on vinyl cap with Teflon liner and screw-on phenolic cap and packaged in specially designed septum-sealed bottles under nitrogen to maintain low water content and ensure product integrity.
CAS Max % 1
UN Number UN1219
Percent Purity Approx.99.5%
Packaging Amber Glass Bottle
Quantity 100 mL