Learn More
Isopropyl Alcohol, AR, For ACS analysis, Macron Fine Chemicals™
macron
Manufacturer:  Avantor Performance Materials MK303206/DEL
Quantity 1L
 View more versions of this product
Catalog No. 14-650-370
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Chemical Identifiers
67-63-0
99.5 % min.
60.1
ACS Reagent Grade
Liquid
CH3CHOHCH3
Isopropyl Alcohol, ACS
Specifications
67-63-0
99.5 % min.
60.1
1L
ACS Reagent Grade
Liquid
CH3CHOHCH3
Clear colorless
Isopropyl Alcohol, ACS