Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade, 99.7%, Spectrum™

missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O

HP692, 67-63-0, C3H8O

Spectrum™ Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Isopropyl Alcohol chromatography solvents are delivered with an expiration date.
CAS Max % 1
UN Number UN1219
Percent Purity 99.7%
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Water 0.0005
Quantity 6 x 1 L