Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, FCC, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O

I1056, 67-63-0, C3H8O

Spectrum™ Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Acidity 10 mg/kg
UN Number UN1219
Percent Purity 99.7%
Lead (Pb) 1 mg/kg
Non_volatile Matter 10 mg/kg
Packaging Amber Glass Bottle
Specific Gravity 0.784
Water 0.002
Quantity 1 L