Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Meets or Exceeds A.C.S. Specifications, SpectroSolv (TM), 99.5%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
67-63-0
67-63-0
CAS Max % 1
Percent Purity 0.995
Grade High Purity Solvent
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Solubility in Water To Pass Test
Water 0.002

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.