Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
Formula Weight 60.1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 12.00°C
Quantity 500mL
Grade Multi-Compendial/USP
Chemical Name or Material Isopropyl Alcohol