Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O

I1048, 67-63-0, C3H8O

Spectrum™ Isopropyl Alcohol, Reagent, ACS is a compound which is colorless, flammable and has a very strong odor. Isopropyl alcohol has many uses, including as an analytical solvent reagent in the laboratory. Other chemical applications include use as a chemical synthesis intermediate, disinfectant, fuel additive and as a biological specimen preservative.
CAS Max % 1
UN Number UN1219
Percent Purity 99.5%
Packaging Drum
Residue after Evaporation 1e-005
Water 0.002
Quantity 200 L