Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, UniSolv(R), 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
67-63-0
67-63-0
CAS Max % 1
Percent Purity 0.999
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 1 ppm
Solubility in Water To Pass Test
Titratable Acid or Base 0.0001 μeq/g
Water 0.0005
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.