Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isopropyl-beta-D-thiogalactoside, High Purity, Spectrum™ Chemical
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
6 x 1 g
6 x 5 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Isopropyl-beta-D-thiogalactoside, High Purity (IPTG) is a compound and a molecular biology reagent. IPTG is an effective inducer of protein expression in the concentration range of 100 μM to 1.5 mM. Spectrum offers highly pure reagents suitable for molecular biology research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Grade | High Purity |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction