Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl cinnamate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
7780-06-5
C12H14O2
190.24
MFCD00048292
RGACABDFLVLVCT-CMDGGOBGSA-N
isopropyl cinnamate,cinnamic acid, isopropyl ester,iso-propyl cinnamate,2-propenoic acid, 3-phenyl-, 1-methylethyl ester,1-methylethyl 3-phenylpropenoate,unii-6rwc10bdd7,isopropyl 3-phenylpropenoate,fema no. 2939,trans-cinnamic acid isopropyl ester,isopropyl 3-phenylacrylate
5273464
propan-2-yl (E)-3-phenylprop-2-enoate
CC(C)OC(=O)C=CC1=CC=CC=C1
7780-06-5
C12H14O2
190.24
MFCD00048292
RGACABDFLVLVCT-CMDGGOBGSA-N
isopropyl cinnamate,cinnamic acid, isopropyl ester,iso-propyl cinnamate,2-propenoic acid, 3-phenyl-, 1-methylethyl ester,1-methylethyl 3-phenylpropenoate,unii-6rwc10bdd7,isopropyl 3-phenylpropenoate,fema no. 2939,trans-cinnamic acid isopropyl ester,isopropyl 3-phenylacrylate
5273464
propan-2-yl (E)-3-phenylprop-2-enoate
CC(C)OC(=O)C=CC1=CC=CC=C1
Quantity 10g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula C6H5CHCHCOOCH(CH3)2
Formula Weight 190.24
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Isopropyl cinnamate
Show More Show Less