Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isopropyl cyanoacetate, 97%

Manufacturer:  Alfa Aesar™ A1188714

Quantity 25g

 View more versions of this product

Catalog No. AAA1188714


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

13361-30-3
127.14
1756014
Isopropyl cyanoacetate
NCCH2CO2CH(CH3)2
MFCD00019842
Cyanoacetic acid isopropyl ester

Specifications

92°C (198°F)
25g
NCCH2CO2CH(CH3)2
MFCD00019842
Cyanoacetic acid isopropyl ester
97%
207°C
13361-30-3
127.14
1756014
Isopropyl cyanoacetate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AG4270000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit