Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl ether, 99+%, extra pure, stabilized with BHT, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass bottle
Metal drum
108-20-3,128-37-0
C6H14O
102.177
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
108-20-3,128-37-0
C6H14O
102.177
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
Water 0.1% max.
Refractive Index 1.3670 to 1.3690 (20°C, 589nm)
Acidity 0.005% max.
Infrared Spectrum Authentic
Residue after Evaporation 0.01% max.
Vapor Pressure 180mbar at 20°C
Quantity 25L
Stabilizer 5 to 250ppm BHT
Packaging Metal drum
Color Undesignated
Density 0.7200g/mL
Assay Percent Range 99+%
Linear Formula [(CH3)2CH]2O
UN Number 1159
Beilstein 01, 362
Merck Index 15, 5258
Formula Weight 102.18
Physical Form Liquid
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Isopropyl ether, Stabilized with BHT
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Danger