Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl ether, for analysis, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
108-20-3,128-37-0
C6H14O
102.18
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
108-20-3,128-37-0
C6H14O
102.18
MFCD00008880
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
diisopropyl ether,isopropyl ether,propane, 2,2'-oxybis,2-isopropoxypropane,diisopropyl oxide,diisopropylether,ether, isopropyl,2,2'-oxydipropane,isopropylether,bis isopropyl ether
7914
2-propan-2-yloxypropane
CC(C)OC(C)C
For analysis
Melting Point -85.5°C
Flash Point -29°C
Refractive Index 1.3670 to 1.369
Viscosity 0.38 mPa.s (25°C)
Specific Gravity 0.72
Boiling Point 68°C
Water 0.1% max.
Density 0.7200g/mL
Packaging Glass bottle
Stabilizer aprox. 50ppm BHT
Acidity 0.0004 meq/g max.
Infrared Spectrum Authentic
Quantity 1L
Color Undesignated
Linear Formula [(CH3)2CH]2O
Merck Index 15, 5258
Solubility Information Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 102.18
Physical Form Liquid
Percent Purity ≥99.0%
Grade Analysis
Chemical Name or Material Isopropyl ether
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May form explosive peroxides.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-560-6