Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl myristate, 98%, Alfa Aesar™

Click to view available options
Quantity:
100mL
500mL
2500mL
110-27-0
C17H34O2
270.457
MFCD00008982
AXISYYRBXTVTFY-UHFFFAOYSA-N
isopropyl myristate,isopropyl tetradecanoate,estergel,bisomel,isomyst,promyr,tetradecanoic acid, 1-methylethyl ester,deltyl extra,kesscomir,tegester
8042
propan-2-yl tetradecanoate
CCCCCCCCCCCCCC(=O)OC(C)C
110-27-0
C17H34O2
270.457
MFCD00008982
AXISYYRBXTVTFY-UHFFFAOYSA-N
isopropyl myristate,isopropyl tetradecanoate,estergel,bisomel,isomyst,promyr,tetradecanoic acid, 1-methylethyl ester,deltyl extra,kesscomir,tegester
8042
propan-2-yl tetradecanoate
CCCCCCCCCCCCCC(=O)OC(C)C
Applications
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

Solubility
Miscible with alcohol. Immiscible with water and glycerol.

Notes
Incompatible with strong oxidizing agents.
Density 0.85 g/mL
Flash Point 152°C (306°F)
Boiling Point 191°C to 194°C (20 mmHg)
Quantity 2500mL
Assay Percent Range 0.98
Solubility Information Miscible with alcohol. Immiscible with water and glycerol.
Formula Weight 270.46
Physical Form Liquid
Chemical Name or Material Isopropyl Myristate

EINECSNumber : 203-751-4

TSCA : Yes

Recommended Storage : Ambient temperatures