Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Myristate, NF, 90%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
110-27-0
C17H34O2
270.46
AXISYYRBXTVTFY-UHFFFAOYSA-N
propan-2-yl tetradecanoate
CCCCCCCCCCCCCC(=O)OC(C)C
110-27-0
C17H34O2
270.46
AXISYYRBXTVTFY-UHFFFAOYSA-N
propan-2-yl tetradecanoate
CCCCCCCCCCCCCC(=O)OC(C)C

IS145, 110-27-0, C17H34O2

Spectrum™ Isopropyl Myristate, NF is an emollient ester of low viscosity used to reduce the greasiness of emulsions. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Drum
Ignition Residue 0.001
Refractive Index 1.432-1.436
Quantity 200 L
CAS Max % 1
Percent Purity 90%