Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isopropyl N-phenylcarbamate, 96%

Click to view available options
Quantity:
5g
25g
100g
122-42-9
C10H13NO2
179.219
MFCD00026382
VXPLXMJHHKHSOA-UHFFFAOYSA-N
propham,isopropyl n-phenylcarbamate,isopropyl phenylcarbamate,triherbide,agermin,collavin,prophame,tuberit,tuberite,birgin
24685
CHEBI:82035
propan-2-yl N-phenylcarbamate
CC(C)OC(=O)NC1=CC=CC=C1
122-42-9
C10H13NO2
179.219
MFCD00026382
VXPLXMJHHKHSOA-UHFFFAOYSA-N
propham,isopropyl n-phenylcarbamate,isopropyl phenylcarbamate,triherbide,agermin,collavin,prophame,tuberit,tuberite,birgin
24685
CHEBI:82035
propan-2-yl N-phenylcarbamate
CC(C)OC(=O)NC1=CC=CC=C1
Melting Point 86°C to 89°C
Quantity 5g
Formula Weight 179.22
Percent Purity 96%
Chemical Name or Material Isopropyl N-phenylcarbamate

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : FD9100000

TSCA : TSCA