Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Palmitate, NF, 90%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
142-91-6
C19H38O2
298.51
XUGNVMKQXJXZCD-UHFFFAOYSA-N
propan-2-yl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OC(C)C
142-91-6
C19H38O2
298.51
XUGNVMKQXJXZCD-UHFFFAOYSA-N
propan-2-yl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OC(C)C

IS160, 142-91-6, C19H38O2

Spectrum™ Isopropyl Palmitate, NF is derived from palm oil and is used as a thickening agent and antistatic agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Refractive Index 1.435-1.438
Quantity 500 mL
CAS Max % 1
Percent Purity 90%