Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Cubitainer
Poly Bottle
Poly Drum
Steel Drum
Description
Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 70%, 30% |
UN Number | UN1219 |
Percent Purity | 68 to 72% |
Grade | USP |
Quantity | 4 L |
Packaging | Poly Bottle |
Specific Gravity | 0.872-0.883 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction