Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Cubitainer
Poly Bottle
Poly Drum
Steel Drum
67-63-0,7732-18-5
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O
67-63-0,7732-18-5
C3H8O
60.10
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
CC(C)O

IS120, 67-63-0, C3H8O

Spectrum™ Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 0.7,0.3
UN Number UN1219
Percent Purity 68 to 72%
Non_volatile Matter 0.0001
Packaging Poly Drum
Specific Gravity 0.872-0.883
Quantity 20 L