Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylbarbituric acid, 97%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
7391-69-7
C7H10N2O3
170.168
GFPIGNBQTXNNAG-UHFFFAOYSA-N
5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
81874
5-propan-2-yl-1,3-diazinane-2,4,6-trione
CC(C)C1C(=O)NC(=O)NC1=O
7391-69-7
C7H10N2O3
170.168
GFPIGNBQTXNNAG-UHFFFAOYSA-N
5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
81874
5-propan-2-yl-1,3-diazinane-2,4,6-trione
CC(C)C1C(=O)NC(=O)NC1=O
Melting Point 215.0°C to 217.0°C
Quantity 25g
Formula Weight 170.17
Chemical Name or Material Isopropylbarbituric acid

RUO – Research Use Only