Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Isopropylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
Description
Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
Linear Formula | (CH3)2CHMgCl |
Quantity | 100 mL |
UN Number | UN3399 |
Sensitivity | Air and moisture sensitive |
Solubility Information | It reacts violently with water. |
Formula Weight | 102.85 |
Concentration or Composition (by Analyte or Components) | 1M in MeTHF |
Chemical Name or Material | Isopropylmagnesium chloride |
Safety and Handling
Hazard Category | H225-H260-H302-H314-H335 |
Hazard Statement | GHS H Statement H225-H261-H314-H318 Highly flammable liquid and vapor. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction