Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
1068-55-9,109-99-9
C3H7ClMg
102.844
MFCD00000466
IUYHWZFSGMZEOG-UHFFFAOYSA-M
isopropylmagnesium chloride,magnesium, chloro 1-methylethyl,chloro 1-methylethyl magnesium,i-prmgcl,isopropylmagnesiumchloride,chloro isopropyl magnesium,isopropyl magnesium chloride,isopropylmagnesium chloride solution,iprmgcl,ipr-mgcl
101955
magnesium;propane;chloride
C[CH-]C.[Mg+2].[Cl-]
1068-55-9,109-99-9
C3H7ClMg
102.844
MFCD00000466
IUYHWZFSGMZEOG-UHFFFAOYSA-M
isopropylmagnesium chloride,magnesium, chloro 1-methylethyl,chloro 1-methylethyl magnesium,i-prmgcl,isopropylmagnesiumchloride,chloro isopropyl magnesium,isopropyl magnesium chloride,isopropylmagnesium chloride solution,iprmgcl,ipr-mgcl
101955
magnesium;propane;chloride
C[CH-]C.[Mg+2].[Cl-]
Linear Formula (CH3)2CHMgCl
Concentration 1.80 to 2.20M
Density 0.9700g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 102.85
Physical Form Solution
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 0.97
Boiling Point 62.4°C (899.8hPa)
Color Brown to Brown-Yellow
Melting Point 5.0°C
Quantity 800mL
Flash Point −27°C
Chemical Name or Material Isopropylmagnesium chloride, 2.0M Solution in THF
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. May cause respiratory irritation. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger