Alfa Aesar™ Isopropylphosphonic dichloride, tech. 90%

Manufacturer: Alfa Aesar™ H2710906

Quantity 5g

 View more versions of this product

Catalog No. AAH2710906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1498-46-0
    Molecular Formula C3H7Cl2OP
    Formula Weight 160.97
    MDL Number MFCD00603000
    UN Number UN3265
    Chemical Name or Material Isopropylphosphonic dichloride
    Grade Technical Grade

    Specifications

    Formula Weight 160.97
    Molecular Formula C3H7Cl2OP
    MDL Number MFCD00603000
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    CAS 1498-46-0
    UN Number UN3265
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Isopropylphosphonic dichloride
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    Isopropylphosphonic dichloride is used as a raw material for the preparation of bis(fluoroalkyl) alkylphosphonates by reacting with fluoroalcohols in the presence of triethylamine as catalyst.

    Solubility
    Miscible with ethylene glycol and diethyl ether.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents, water and bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit