Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyltriphenylphosphonium Iodide 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
24470-78-8
C21H22IP
432.27
MFCD00031595
HHBXWXJLQYJJBW-UHFFFAOYSA-M
isopropyltriphenylphosphonium iodide,phosphonium, 1-methylethyl triphenyl-, iodide,isopropyltriphenylphosphanium iodide,isopropyl triphenyl phosphonium iodide,triphenyl propan-2-yl phosphanium iodide,i-propyl triphenylphosphonium iodide,isopropyl triphenylphosphonium iodide,acmc-209gck,ksc493k7j,isopropyltriphenylphosphine iodide
2723783
triphenyl(propan-2-yl)phosphanium;iodide
CC(C)[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
24470-78-8
C21H22IP
432.27
MFCD00031595
HHBXWXJLQYJJBW-UHFFFAOYSA-M
isopropyltriphenylphosphonium iodide,phosphonium, 1-methylethyl triphenyl-, iodide,isopropyltriphenylphosphanium iodide,isopropyl triphenyl phosphonium iodide,triphenyl propan-2-yl phosphanium iodide,i-propyl triphenylphosphonium iodide,isopropyl triphenylphosphonium iodide,acmc-209gck,ksc493k7j,isopropyltriphenylphosphine iodide
2723783
triphenyl(propan-2-yl)phosphanium;iodide
CC(C)[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]
For C-C Bond Formation (Olefination)
Packaging Glass bottle
Color Beige-Brown to Yellow
Quantity 100g
Infrared Spectrum Authentic
Melting Point 192.0°C to 197.0°C
Assay Percent Range 98%
Linear Formula (CH3)2CHPPh3+I-
Formula Weight 432.27
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Isopropyltriphenylphosphonium iodide, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 246-281-5