Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-N-oxide, 94%, Acros Organics

Click to view available options
Quantity:
25g
1532-72-5
C9H7NO
145.161
MFCD00006903
RZIAABRFQASVSW-UHFFFAOYSA-N
isoquinoline n-oxide,isoquinoline 2-oxide,isoquinoline-n-oxide,isoquinoline oxide,isoquinoline, 2-oxide,isoquinolin-n-oxide,isoquinolin-2-ium-2-olate,isoquinolinen-oxide,acmc-1boxv,2-oxido-isoquinolin-2-ium
290378
2-oxidoisoquinolin-2-ium
C1=CC=C2C=[N+](C=CC2=C1)[O-]
1532-72-5
C9H7NO
145.161
MFCD00006903
RZIAABRFQASVSW-UHFFFAOYSA-N
isoquinoline n-oxide,isoquinoline 2-oxide,isoquinoline-n-oxide,isoquinoline oxide,isoquinoline, 2-oxide,isoquinolin-n-oxide,isoquinolin-2-ium-2-olate,isoquinolinen-oxide,acmc-1boxv,2-oxido-isoquinolin-2-ium
290378
2-oxidoisoquinolin-2-ium
C1=CC=C2C=[N+](C=CC2=C1)[O-]
Melting Point 103°C to 108°C
Infrared Spectrum Authentic
Quantity 25g
Color Brown to Cream
Assay Percent Range 94%
Beilstein 20, II, 237
Formula Weight 145.16
Physical Form Powder or Chunks
Percent Purity ≥92%
Chemical Name or Material Isoquinoline-N-oxide