Learn More
Fisherbrand™ Isotemp CO2 Incubator Available on GSA/VA Contract for Federal Government customers only.
Value for everyday cell culturing . The Fisherbrand CO2 incubator provides proven technologies for reliable, homogenous growth environment and dependable quality.
$7380.00 - $8770.00
Specifications
Frequency 50/60 Hz
Chamber Material Stainless Steel
Capacity (Metric) 150 L
View More Specs
Products 2
Catalog Number CO2 Sensor Technology Price Quantity & Availability  
Catalog Number CO2 Sensor Technology Price Quantity & Availability  
11-676-604 Available on GSA/VA Contract for Federal Government customers only. Accessories View Documents Thermal Conductivity (TC) Each for $7,380.00
Add to cart
11-676-605 Available on GSA/VA Contract for Federal Government customers only. Accessories View Documents Infrared (IR) Each for $8,770.00
Add to cart
Description
Description
  • Efficient high performance heaters surround the chamber for even temperature distribution
  • Gentle fan assisted air flow pattern is designed for uniform distribution of temperature, CO2, and humidity
  • The 90°C high temperature decontamination cycle simplifies cleaning because no autoclave and toxic chemicals required
  • The cycle eliminates variability in disinfection and has been proven effective against wide range of contaminants including bacteria, molds, and fungal spores and mycoplasma, eliminating the variability in disinfection
  • CO2 sensors are positioned directly in the chamber for precise control. A choice of CO2 sensor technology includes.  
    • Thermal Conductivity (TC) for precise monitoring and reliable long life
    • Infrared (IR) for independent operation for precise, remote monitoring applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
50/60 Hz
Stainless Steel
150 L