Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Explosion Proof Under Counter Freezer Available on GSA/VA Contract for Federal Government customers only.

For storing volatile materials in areas with explosive atmospheres or in hazardous locations

$3550.00

Specifications

Defrost Manual
View More Specs
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Products 2
Catalog Number Description Capacity (English) Certifications/Compliance Dimensions (L x W x H) Exterior Price Quantity & Availability  
Catalog Number Description Capacity (English) Certifications/Compliance Dimensions (L x W x H) Exterior Price Quantity & Availability  
05EFEEFSA Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Undercounter 5 cu. ft. NFPA, OSHA, UL 29.75 x 21 x 33.5 in. (74.9 x 54.6 x 85.1 cm)
Each for $3,550.00
Add to cart
 
20EFEEFSA
Accessories View Documents
Upright 20.9 cu. ft. cCSAus 39.61 x 33 x 67.25 in. (100.6 x 83.8 x 170.8 cm) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Temperature range: -20° to -12°C
  • Adjust setpoint while running
  • Capacities: 20 cu. ft. (566.34L)
  • Manual defrost
  •  
Safety Features
  • Red warning label on front: “Explosion-Proof Freezer” Compressors have enclosed overloads, relays
  • Top-mounted, explosion-proof thermostats
  • Static hot-wall condensers and evaporators are free of electric fans
Construction
  • White, baked-on enameled steel exteriors
  • Hinged solid door
  • White enameled steel interiors
  • R134A, CFC Free refrigerant, polyurethane foam insulation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Manual
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit