Learn More
Fisherbrand™ Isotemp™ Explosion-Proof Refrigerator
For storing volatile materials in areas with explosive atmospheres or in hazardous locations
 View more versions of this product
Catalog No. 05EREEFSA
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Description
Description
Prevent the triggering of an explosion inside or outside the unit. Fisher Scientific™ Isotemp™ Explosion-Proof Refrigerator is suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.
  • Hydraulic thermostat
  • Manual defrost
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and refrigerant
Specifications
Specifications
1°C to 12°C
5.5 cu. ft.
120 V 60 Hz
60 Hz
Not Provided
17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
45.3 kg
54.5 kg
ABS Plastic
13 Months Parts and Labor
Enameled Steel
Manual
Hinged, Open Left to Right, Reversible
Polyurethane Foam
160 w
NFPA 45 and 70, UL
155.7 L
120 V
1.33 A
28.1 x 23.63 x 33.5 in. (71.37 x 64.8 x 85.1 cm)
100 lb.
120 lb.
R134a, 100% CFC Free
Door Lock
White
2 Adjustable Feet In Front, 2 Fixed Pads In Rear
Mechanical / Outside, Upper Rear In Grey Housing
Bottom Mounted
1 (1 compartment)
3 Adjustable Shelves, Storage On Bottom Of Unit
SDS
Documents
Product Certifications
Accessories