Learn More
Fisherbrand™ Isotemp Flammable Materials Freezer Available on GSA/VA Contract for Federal Government customers only.
For storing volatile materials, pharmaceuticals, clinical reagents, and biological samples.
$3110.00
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Products 2
Catalog Number Capacity (English) Price Quantity & Availability  
Catalog Number Capacity (English) Price Quantity & Availability  
05FFEEFSA Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
5 cu. ft.
Each for $3,110.00
Add to cart
20FFEEFSA
Accessories View Documents
20.9 cu. ft. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Accidental ignition of refrigerator contents is prevented by specially designed compressor with enclosed overload, relay, and hydraulic thermostat
  • Static condenser free of electric fan
  • Aluminum evaporator coil
  • Two fixed shelves, three door shelves
  • Requires extra clearance when recessed
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.