Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ General Purpose Chromatography Refrigerators, Sliding Glass Door

Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand™ Isotemp™ Chromatography Refrigerators for General Purpose Use are designed with your needs in mind.

Manufacturer:  Fisherbrand™ FBG45CSLA

 View more versions of this product

Catalog No. FBG45CSLA


Add to cart

Description

Description

  • Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
  • Whisper-quiet operation
  • ENERGY STAR certified
  • Temperature, door ajar, and power failure (with battery back-up) alarms
  • Green, natural refrigerants
  • Adaptive defrost
  • Glass (double pane) or solid door configurations
  • Choice of white painted interior and exterior, or stainless steel interior and gray exterior
  • Digital controller
  • Rear access port
  • Remote alarm contacts
  • Keyed door lock(s)
  • LED interior lighting
  • Low heat output
Specifications

Specifications

100 lb./full shelf; 50 lb./half shelf
Key Lock
Gray
45 cu. ft.
2
27 x 49 x 61.75 in. (686 x 1245 x 1569 mm)
2°C to 10°C
Stainless steel
5°C
CSLA
R290
561 lb.
Sliding Glass Door
45 kg/full shelf; 22.5 kg/half shelf
cULus, Energy Star
Microprocessor
1274 L
32 x 52.7 x 81 in.(813 x 1339 x 813 mm)
NEMA 5-15 P
115 V/60 Hz
2 years parts and labor, plus 3 year compressor warranty
5 A
1 Rear access port and 2 side access ports
8 full shelves plus 2 half shelves
255 kg
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit